Masterflex® Smart Gasket® Sanitary Sampling Gasket, Viton, 1 Port, 1-1/2" Tri-Clamp; 1/EA
Katalogové číslo MFLX30542-14
Dodavatel: Avantor Fluid Handling
O tomto produktu
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):1-1/2"
- Typ připojení:Sanitary
- Popis:Sanitary Sampling Gasket, Viton, 1 Port, 1-1/2" Tri-Clamp; 1/EA
- Sekvence:33
- Environmentally Preferable:
- Bal.:1
- Fitting material:Viton®
- Sanitary gasket size:1-1/2"
- No. of ports:1
- Záruka:1 year
Frequently Bought Together