Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
25, Sanitary Sampling Gasket, Silicone, 1 Port, 3/4" Tri-Clamp; 1/EA, Platinum-Cured Silicone, 3/4", 3/4"
Katalogové číslo MFLX30542-04
Dodavatel: Avantor Fluid Handling
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Katalogové číslo MFLX30542-04
Dodavatel: Avantor Fluid Handling
Přejít na:
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Certifikáty
Hledat podle čísla šaržeSpecifikace
- Sanitary clamp size (Port 1):3/4"
- Popis:Sanitary Sampling Gasket, Silicone, 1 Port, 3/4" Tri-Clamp; 1/EA
- Sekvence:25
- Fitting material:Platinum-Cured Silicone
- Sanitary gasket size:3/4"
- Cat. No.:MFLX30542-04
- Typ připojení:Sanitary
- Záruka:1 year
- No. of ports:1