Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/4" ID x 5/32" ID; 100/PK
Katalogové číslo MFLX40626-26
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
- Connection Type: 1/4" ID x 5/32" ID
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Velikost ozubu/vnit. Ø hadičky (port 1):½"
- Velikost ozubu/vnit. Ø hadičky (port 2):⁵⁄₃₂"
- Typ připojení:Barbed Reducer
- Flow pattern:Straight
- Popis:Fitting, polypropylene, straight, hose barb reducer, int.Ø ½×⁵⁄₃₂";10/pack
- Sekvence:29
- Environmentally Preferable:
- Model:40626-26
- Sterilizace:Ethylene oxide (ETO) or gamma
- Bal.:100
- Fitting material:Polypropylene
Frequently Bought Together