Masterflex® Fitting, Polypropylene, Tee, Hose Barb Union 3/16" ID; 25/PK
Katalogové číslo MFLX40623-70
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Velikost ozubu/vnit. Ø hadičky (port 1):³⁄₁₆"
- Velikost ozubu/vnit. Ø hadičky (port 2):³⁄₁₆"
- Typ připojení:Barbed Union
- Flow pattern:T
- Popis:Fitting, Polypropylene, Tee, Hose Barb Union ³⁄₁₆" ID; 25/PK
- Sekvence:30
- Environmentally Preferable:
- Model:40623-70
- Orifice dimensions:0,09375"
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Bal.:25
- Barb size/tubing int.Ø (port 3):³⁄₁₆"
- Fitting material:Polypropylene
Frequently Bought Together