Masterflex® Fitting, Polypropylene, Straight, Hosebarb to Threads, 1/2" ID x 1/2" NPT(M); 10/PK
Catalog # MFLX40610-46
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/2" ID x 1/2" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Velikost ozubu/vnit. Ø hadičky (port 2):1/2"
- Typ připojení:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/2"
- Popis:Fitting, Polypropylene, Straight, Hosebarb to Threads, 1/2" ID x 1/2" NPT(M); 10/PK
- Sekvence:27
- Environmentally Preferable:
- Model:40610-46
- No. of ports:2
- Orifice dimensions:8,73 mm (0,34375")
- Sterilizace:Ethylene oxide (ETO) or gamma
- Záruka:Out of box
- Bal.:10
- Fitting material:Polypropylene
Frequently Bought Together