Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 3/4" ID x 1/2" ID; 10/PK
Catalog # MFLX30616-71
Supplier: Avantor Fluid Handling
CE certificate
About this item
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Velikost ozubu/vnit. Ø hadičky (port 1):¾"
- Velikost ozubu/vnit. Ø hadičky (port 2):½"
- Typ připojení:Barbed Reducer
- Flow pattern:Straight
- Popis:Fitting, polypropylene, straight, hose barb reducer, int.Ø ¾×½";10/pack
- Sekvence:04
- Environmentally Preferable:
- Model:30616-71
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Bal.:10
- Fitting material:Polypropylene
Frequently Bought Together