0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
DIFFERENTIATOR FAST ALCOHOL/ACETONE 2.5L 1 * 2,5 L
720-1165
:
RAL
DIFFERENTIATOR FAST ALCOHOL/ACETONE 2.5L 1 * 2,5 L
720-1165
:
RAL
Frequently Bought Together
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Vizmut 1000 mg/L v 0,5 M kyselině dusičné (from Bi(NO₃)₃) Certipur® standard pro AAS, Supelco®
: Merck
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
: Merck
Test pro stanovení volného chloru, MQuant™, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
: Merck
May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
: Merck
Accessories for PIPETBOY acu / pro
: Integra
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
SAFRANIN 1L 1 * 1 L
: RAL