4E-BP1 (TOTAL) AND 4E-BP1 (PHO-THR37/46) 1 * 1 KIT
Katalogové číslo CAYM500225-480
Dodavatel: Cayman Chemical
4E-BP1 (TOTAL) AND 4E-BP1 (PHO-THR37/46) 1 * 1 KIT
Katalogové číslo CAYM500225-480
Dodavatel: Cayman Chemical
Frequently Bought Together
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Cyklohexan, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Ethylacetát ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
Dodavatel: VWR Chemicals
Toluen ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Triethylamin ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Vanilin for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Petrolether, 40…60 °C, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Aceton ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Cyklohexan ≥99.8%, SupraSolv® for GC-MS
Dodavatel: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Dodavatel: VWR Chemicals
n-Hexan, Uvasol® for spectroscopy, Supelco®
Dodavatel: Merck
Ethylendiamin ≥99.0% (by GC), GPR RECTAPUR®
Dodavatel: VWR Chemicals
Tetrachlorethylen 99%, extra pure
Dodavatel: Thermo Fisher Scientific