STAT5 (TOTAL) AND STAT5 (PHO-TYR694/699) 1 * 1 KIT
Katalogové číslo CAYM500217-480
Dodavatel: Cayman Chemical
STAT5 (TOTAL) AND STAT5 (PHO-TYR694/699) 1 * 1 KIT
Katalogové číslo CAYM500217-480
Dodavatel: Cayman Chemical
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
1-methyl-2-pyrolidon for peptide synthesis, Supelco®
Dodavatel: Merck
2-Etoxyethanol for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Chlorbenzen ≥99%
Dodavatel: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochlorid, Supelco®
Dodavatel: Merck
Chloroform 99.0-99.4% (by GC) stabilizovaný, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Glycerin solution 86-89% (by titrimetric analysis), Puriss. p.a., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Dodavatel: Merck
Dichlormethan ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Dodavatel: VWR Chemicals
N,N-Dimethylformamid, extra dry over Molecular Sieve 99.8%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Dodavatel: Honeywell Chemicals