Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
13, Plan achromat N.A. 0,65, 40×, 0,8 mm
Catalog # ECHLRMAG04022
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG04022
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
13, Plan achromat N.A. 0,65, 40×, 0,8 mm
Price
5 070,00 Kč
Log in for availability
1 * 1 KS
Specifications
  • Zvětšení:
    40×
  • Popis:
    Plan achromat N.A. 0,65
  • Sekvence:
    13
  • Pracovní vzdálenost:
    0,8 mm