Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
15, Watch glass, glass, with fused rim, 90 mm
Katalogové číslo HECH40010009
Watch glasses
Katalogové číslo HECH40010009
Watch glasses
Watch glasses
15, Watch glass, glass, with fused rim, 90 mm
Přejít na:
Price
516,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341
Specifikace
  • Ø:
    90 mm
  • Popis:
    Watch glass, glass, with fused rim
  • Sekvence:
    15