Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
13, Watch glass, glass, with fused rim, 70 mm
Catalog # HECH40010007
Watch glasses
Catalog # HECH40010007
Watch glasses
Watch glasses
13, Watch glass, glass, with fused rim, 70 mm
Price
501,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341
Specifications
  • Ø:
    70 mm
  • Popis:
    Watch glass, glass, with fused rim
  • Sekvence:
    13