[EN]MEASURE KIT FOR ETHYL GLUCURONIDE(ET 1 * 25 Tests
Katalogové číslo SURES1DSETG1000
Dodavatel: SURESCREEN
[EN]MEASURE KIT FOR ETHYL GLUCURONIDE(ET 1 * 25 Tests
Katalogové číslo SURES1DSETG1000
Dodavatel: SURESCREEN
Frequently Bought Together
Test kits, ammonium, Spectroquant®, Supelco®
Dodavatel: Merck
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Dodavatel: MACHEREY-NAGEL
Formaldehyde 4% stabilised, buffered (pH 7,0 ±0,1)
Dodavatel: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Dodavatel: Merck
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
VWR®, Práškové balíčky pro stanovení chlóru, činidla pro fotometry
Dodavatel: VWR Chemicals
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, chloride, Spectroquant®, Supelco®
Dodavatel: Merck
VWR®, Kryoboxy, lepenka, 133×133 mm
Dodavatel: VWR Collection
Kyselina chlorovodíková 1 mol/l (1 N) for biopharmaceutical production
Dodavatel: VWR Chemicals
Formalin 10%, Q Path®, buffered
Dodavatel: VWR Chemicals
Drug Test, 6 Parameter Multi-Panel Dip Urine
Dodavatel: SURESCREEN