Rigid threaded couplings, BOLA
03, Threaded couplings, 24 mm distance between sealing lips, 51 mm, GL 25
Katalogové číslo BOHLH903-25
Dodavatel: Bohlender
Přejít na:
Product Details & Documents
Special design allows turning the two halfs of the coupling independently from each other, and to adjust connected components. This design also provides high stability. A sealing lip on the connecting tube with integrated FKM O-ring ensures good sealing on the GL thread.
- Sturdy coupling with female GL threads made of PPS
- Internal connecting tube made of PTFE
- Universal chemical resistance
- Flowing product is only exposed to PTFE
For connecting two GL threads of the same size.
Specifikace
- Závit:GL 25
- Popis:Threaded couplings, 24 mm distance between sealing lips
- Délka:51 mm
- Sekvence:03



