Rigid threaded couplings, BOLA
01, Threaded couplings, 28 mm distance between sealing lips, 49 mm, GL 14
Catalog # BOHLH903-14
Supplier: Bohlender
Jump to:
Product Details & Documents
Special design allows turning the two halfs of the coupling independently from each other, and to adjust connected components. This design also provides high stability. A sealing lip on the connecting tube with integrated FKM O-ring ensures good sealing on the GL thread.
- Sturdy coupling with female GL threads made of PPS
- Internal connecting tube made of PTFE
- Universal chemical resistance
- Flowing product is only exposed to PTFE
For connecting two GL threads of the same size.
Specifications
- Závit:GL 14
- Popis:Threaded couplings, 28 mm distance between sealing lips
- Délka:49 mm
- Sekvence:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



