[EN]1-PROPOXY-2-PROPANOL (CONTAINS 2-ISO 1 * 25 mL
Katalogové číslo ABCRAB474676-25ML
Dodavatel: abcr
[EN]1-PROPOXY-2-PROPANOL (CONTAINS 2-ISO 1 * 25 mL
Katalogové číslo ABCRAB474676-25ML
Dodavatel: abcr
Frequently Bought Together
Toluen 99.8+% p.a.
Dodavatel: Thermo Fisher Scientific
Chloroform ≥99.5% stabilizovaný
Dodavatel: Honeywell Chemicals
Butanon 99+% for spectroscopy
Dodavatel: Thermo Fisher Scientific
Benzylalkohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
COENZYME Q-10, 98+% 1 * 5 g
Dodavatel: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Ethylacetát ≥99.5% for HPLC
Dodavatel: Thermo Fisher Scientific
n-Hexan ≥95% for spectrophotometry
Dodavatel: Thermo Fisher Scientific
[EN]OXSILAN ICP STANDARD 1 - 11 ELEMENTS 1 * 1 L
Dodavatel: CUSTOM MADE CHEMICALS LAB
N,N-Dimethylacetamid ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
2-methylbutan ≥99%, Reagent Grade
Dodavatel: Honeywell Chemicals
2-Butoxyethanol ≥99%
Dodavatel: Thermo Fisher Scientific
Dichlormethan 99.8% stabilised for HPLC
Dodavatel: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Dodavatel: Thermo Fisher Scientific