Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Chlorine dioxide
1, QUANTOFIX Chlorine dioxide 15, 0.25 • 0.5 • 1 • 3 • 5 • 10 • 15 - mg/L, Colour reaction with a triazine derivate, 40×40×120 mm (package), 50 test(s)
Catalog # MANA91355
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Chlorine dioxide
Catalog # MANA91355
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Chlorine dioxide
QUANTOFIX® Test Strips, Chlorine dioxide
1, QUANTOFIX Chlorine dioxide 15, 0.25 • 0.5 • 1 • 3 • 5 • 10 • 15 - mg/L, Colour reaction with a triazine derivate, 40×40×120 mm (package), 50 test(s)
CAS Number:  
Price
679,00 Kč
Log in for availability
1 * 50 Tests
Product Details & Documents
Semi-quantitative test strips for the determination of chlorine dioxide.
  • Chlorine dioxide - 0, 10, 25, 50, 100, 250, 500, 1000, 1500, 2000, 2500, 3000 mg/L
  • Chlorine dioxide - Yellow → dark green (field 1), colourless → brown-black (field 2)
Ideal for rapid and easy analysis of chlorine dioxide directly on-site. No maintenance or calibration is required.
Specifications
Specifications
  • Chemická metoda:
    Colour reaction with a triazine derivate
  • Popis:
    QUANTOFIX Chlorine dioxide 15
  • Rozměry:
    40×40×120 mm (package)
  • Sekvence:
    1
  • Testů v kitu:
    50 test(s)
  • Rozsah:
    0.25 • 0.5 • 1 • 3 • 5 • 10 • 15 - mg/L
  • Obsah:
    50 test strips in a tube, instruction leaflet
  • Hmotnost:
    35 g
  • Cat. No.:
    MANA91355
  • Plošina:
    Semi-quantitative test strips
  • Parametry:
    All reagents included
  • Přístroj:
    No
  • Životnost police (měsíce):
    24 (from production)
  • Shoda:
    No hazardous material
  • Metoda:
    Chlorine dioxide - Redox reaction (colorless → pink)
Specifications