Orders LinkContactUsLinkComponent
Wire baskets, round, with handle
Wire baskets, round, with handle
02, Wire basket, round, with handle, 160×180 mm
Katalogové číslo 216-2377
Dodavatel:   RSG
Wire baskets, round, with handle
Katalogové číslo 216-2377
Dodavatel:  RSG
Wire baskets, round, with handle
Wire baskets, round, with handle
02, Wire basket, round, with handle, 160×180 mm
Přejít na:
Price
5 600,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Made from stainless steel 1.4301.
  • Rust-proof
  • Very good temperature resistance
  • Good chemical resistance
  • With reinforced edges
Specifikace
  • Ø×H:
    160×180 mm
  • Popis:
    Wire basket, round, with handle
  • Sekvence:
    02