Orders LinkContactUsLinkComponent
Wire baskets, round, with handle
Wire baskets, round, with handle
01, Wire basket, round, with handle, 100×80 mm
Catalog # 216-2376
Supplier:   RSG
Wire baskets, round, with handle
Catalog # 216-2376
Supplier:  RSG
Wire baskets, round, with handle
Wire baskets, round, with handle
01, Wire basket, round, with handle, 100×80 mm
Price
2 000,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Made from stainless steel 1.4301.
  • Rust-proof
  • Very good temperature resistance
  • Good chemical resistance
  • With reinforced edges
Specifications
  • Ø×H:
    100×80 mm
  • Popis:
    Wire basket, round, with handle
  • Sekvence:
    01