Orders LinkContactUsLinkComponent
FUCHSIN BASIC CERTIFIED 1 * 10 g
FUCHSIN BASIC CERTIFIED 1 * 10 g
Catalog # HIMERM112-10G
Supplier:   HIMEDIA
FUCHSIN BASIC CERTIFIED 1 * 10 g
Catalog # HIMERM112-10G
Supplier:  HIMEDIA
CAS Number:  
FUCHSIN BASIC CERTIFIED 1 * 10 g
CAS Number:  
Price
601,00 Kč
Log in for availability
1 * 10 g