Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Picolinamide 95%

Picolinamide 95%

3173, Bottle, 5 g
Catalog # APOSOR937776-5G
Supplier:   Apollo Scientific
Picolinamide 95%
Catalog # APOSOR937776-5G
Supplier:  Apollo Scientific
CAS Number:  1452-77-3
Picolinamide 95%

3173, Bottle, 5 g

CAS Number:  1452-77-3
Price
697,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Pyridinecarboxamide, Pyridine-2-carboxamide
  • CAS Number:1452-77-3
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00023483
  • Molecular Weight:122.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    5 g
  • Sekvence:
    3173

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.