Orders LinkContactUsLinkComponent
Laboratory software, Labworldsoft 6
Laboratory software, Labworldsoft 6
03, Labworldsoft® 6 Visc, Viscosity measurements for thermostats and pH meters of all manufacturers
Catalog # IKAA20101872
Supplier:   IKA
Laboratory software, Labworldsoft 6
Catalog # IKAA20101872
Supplier:  IKA
Laboratory software, Labworldsoft 6
Laboratory software, Labworldsoft 6
03, Labworldsoft® 6 Visc, Viscosity measurements for thermostats and pH meters of all manufacturers
Price
32 810,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Labworldsoft 6 is a laboratory software to control lab devices, automate processes, and to document your measurements and results. The software provides access to a huge library of different laboratory devices.
  • Easy recording of various physical parameters
  • Function modules to create time triggered or event triggered configurations
  • Communication through RS232, USB, Bluetooth or network
  • Supports formats .csv, .xslx and ASCII
  • Compatibility with Windows 7, 8 and 10
Labworldsoft does not integrate only IKA laboratory devices, but supports several other device from different manufacturers as well. You stay flexible in the choice of your lab devices you want to use and profit from the whole device range of the laboratory world.
Delivery information: Includes one year of support with new versions of Labworldsoft 6.
Specifications
  • Popis:
    Labworldsoft® 6 Visc
  • Sekvence:
    03
  • Určeno pro:
    Viscosity measurements for thermostats and pH meters of all manufacturers