Orders LinkContactUsLinkComponent
Samplers, PowderProof®
Samplers, PowderProof®
10, Spiral screw for PowderProof® for powders up to 2 mm Ø, for 600 mm
Katalogové číslo BURK5302-0616
Dodavatel:   BURKLE
Samplers, PowderProof®
Katalogové číslo BURK5302-0616
Dodavatel:  BURKLE
Samplers, PowderProof®
Samplers, PowderProof®
10, Spiral screw for PowderProof® for powders up to 2 mm Ø, for 600 mm
Price
11 290,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Quality assurance starts with the inspection of incoming goods. An all-layer sample of bulk goods is no longer a problem with PowderProof®, the sampler for powders and granules.
  • Many uses
  • Stainless steel AISI 316L (1.4404)/AISI 301 (1.4310)
  • Includes motor, two rechargeable batteries, fast charger, lance, spiral screw, bags, bag clip, and cleaning brush
  • Additional spiral screws available as accessory
PowderProof® is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without any problem. This modular design makes PowderProof® a versatile instrument for sampling powders and granules from drums, barrels, bags, or sacks.
Safe and reliable sampling is made even easier with useful accessories such as basket-enclosed bottles, sample bags, and close-it control seals.
PowderProof® with lance for horizontal use, such as sampling from sacks, with lance for vertical use, for sampling from open containers such as barrels, drums, or bags.
Specifikace
  • Popis:
    Spiral screw for PowderProof® for powders up to 2 mm Ø, for 600 mm
  • Sekvence:
    10
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Transport case
    Transport case
    312-1141
    07
    Přihlásit se pro dostupnost
    1 * 1 KS - 4 130,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Kapacita
    Závit
    Dostupnost
    Price
  • 300-0174
    05
    500 ml
    50 mm
    Přihlásit se pro dostupnost
    1 * 1 KS - 119,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Packaging bags with closure (120×170 mm), 400 ml
    Packaging bags with closure (120×170 mm), 400 ml
    113-5414
    Přihlásit se pro dostupnost
    1 * 100 KS - 420,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Clamp for sampling bags, Ø 25 mm
    Clamp for sampling bags, Ø 25 mm
    BURK5338-0001
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 615,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Basket for sample bottles 500 ml, Ø 84 mm, including holding ring
    Basket for sample bottles 500 ml, Ø 84 mm, including holding ring
    BURK5302-2055
    02
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 270,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Basket for sample bottles 1000 ml, Ø 100 mm, including holding ring
    Basket for sample bottles 1000 ml, Ø 100 mm, including holding ring
    BURK5302-2105
    03
    Přihlásit se pro dostupnost
    1 * 1 KS - 4 150,00 Kč