n-Hexan for gas chromatography, Burdick & Jackson™
5202, Stainless steel can, 7 L
HONC65747-7L-RC
: Hexyl hydride, Hexanen, Hexane
- :110-54-3
- :H₃C(CH₂)₄CH₃
- :MFCD00009520
- :86.18
- :66…69 °C (1,013 hPa)
- :–95 °C
- :0,660…0.668 g/cm3 (20 °C)
- :Ambient
- :203-777-6
- :1208
- :3,II
- :Density: 0,659 g/cm³ (20 °C)
- Assay (GC):Min. 95.0 %
- Non-volatile matter:Max. 0.0003 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for residue analysis:Max. 5 ng/l
- Typ balení:Stainless steel can
- Bal.:7 L
- Sekvence:5202



