Avantor® Hichrom HI-BASIC, GC Columns
GC Column, Capillary, 0,25 µm, 0,32 mm, 30 m
Katalogové číslo HI38-32-025-30
Dodavatel: Avantor
Přejít na:
Product Details & Documents
Application specific column for basic compouns (e.g. amines).
Max temperature limits may change depending on the film thickness.
Specifikace
- Formát kolony:Capillary
- Tloušťka filmu:0,25 µm
- Ø vnitř.:0,32 mm
- Popis:GC Column
- Délka:30 m
- Cat. No.:HI38-32-025-30
- Crossbond:Stabilised
- Phase composition:Proprietary Phase developed for aliphatic and aromatics Amines
- Phase name:HI-BASIC
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PF-C2Přihlásit se pro dostupnost1 * 10 KS - 2 910,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-100-PF-C5Přihlásit se pro dostupnost1 * 100 KS - 21 210,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PFY-C9Přihlásit se pro dostupnost1 * 10 KS - 25 270,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-1-PFY-C6Přihlásit se pro dostupnost1 * 1 KS - 2 620,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-2-PFY-C7Přihlásit se pro dostupnost1 * 2 KS - 5 230,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-25-PF-C3Přihlásit se pro dostupnost1 * 25 KS - 5 650,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-50-PF-C4Přihlásit se pro dostupnost1 * 50 KS - 10 870,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PF-C1Přihlásit se pro dostupnost1 * 5 KS - 1 950,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PFY-C8Přihlásit se pro dostupnost1 * 5 KS - 12 910,00 Kč