Orders LinkContactUsLinkComponent
Microscope objective lens
Microscope objective lens
42, N PLAN 50×/0,9 NA oil; BF, POL, FLUO contrast; transmitted light, 50×, 0,14 mm
Katalogové číslo LEIM11506321
Dodavatel:   LEICA MICROSYSTEMS
Microscope objective lens
Katalogové číslo LEIM11506321
Dodavatel:  LEICA MICROSYSTEMS
Microscope objective lens
Microscope objective lens
42, N PLAN 50×/0,9 NA oil; BF, POL, FLUO contrast; transmitted light, 50×, 0,14 mm
Přejít na:
Price
42 380,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The objective lens is a critical part of the microscope optics. The microscope objective is positioned near the sample, specimen, or object being observed. It has a very important role in imaging, as it forms the first magnified image of the sample.
  • The objective forms the first magnified image of the sample
The numerical aperture (NA) of the objective indicates its ability to gather light and largely determines the microscope’s resolution, the ability to distinguish fine details of the sample.
Objectives are categorised into performance classes on the basis of their transmission, chromatic correction, planarity, and others. From the point of view of the International Organization of Standardisation (ISO), there are three groups of objective classes differing in quality of chromatic correction (Achromats, Semi-Apochromats and Apochromats).
Specifikace
  • Zvětšení:
    50×
  • Popis:
    N PLAN 50×/0,9 NA oil; BF, POL, FLUO contrast; transmitted light
  • Sekvence:
    42
  • Pracovní vzdálenost:
    0,14 mm