Orders LinkContactUsLinkComponent
Accupod solid samplers
Accupod solid samplers
01, Accupod sampler, non sterile, HDPE, 1 ml
Katalogové číslo 300-0585
Dodavatel:   Sampling Systems
Accupod solid samplers
Katalogové číslo 300-0585
Dodavatel:  Sampling Systems
Accupod solid samplers
Accupod solid samplers
01, Accupod sampler, non sterile, HDPE, 1 ml
Přejít na:
Price
5 440,00 Kč
Přihlásit se pro dostupnost
1 * 50 KS
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
  • Suitable for sampling blend uniformity
  • Next generation in sampling technology
  • Designed to minimise sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Specifikace
  • Objem:
    1 ml
  • Popis:
    Accupod sampler, non sterile
  • Materiál:
    HDPE
  • Sekvence:
    01