Orders LinkContactUsLinkComponent
Cyclic Olefin, Reference Standards
Cyclic Olefin, Reference Standards
Cyclic Olefin Polymer, USP reference standard, Vial, 1 g
Katalogové číslo 1152814.
Dodavatel:   USP
Cyclic Olefin, Reference Standards
Katalogové číslo 1152814.
Dodavatel:  USP
Číslo CAS:  
Cyclic Olefin, Reference Standards
Cyclic Olefin Polymer, USP reference standard, Vial, 1 g
Přejít na:
Číslo CAS:  
Price
9 400,00 Kč
Přihlásit se pro dostupnost
1 * 1 g
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikace
Specifikace
  • Typ balení:
    Vial
  • Bal.:
    1 g
  • Standard:
    USP reference standard
  • Popis:
    Cyclic Olefin Polymer