Orders LinkContactUsLinkComponent
Forceps, disposable
Forceps, disposable
05, Broad tip, straight, -, Large pack
Katalogové číslo 300-0349
Dodavatel:   BURKLE
Forceps, disposable
Katalogové číslo 300-0349
Dodavatel:  BURKLE
Forceps, disposable
Forceps, disposable
05, Broad tip, straight, -, Large pack
Přejít na:
Price
1 150,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
  • PS, white
  • Can be individually packed and sterilised
  • For single use
  • Clean room manufactured (Class 7 cleanroom)
  • Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. Light plastic forceps for single use allow for fatigue-free work. Cleanroom manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined.
These lab forceps are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses.
Pointed/angled shape: These disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends.
Pointed shape: These anatomical forceps have a straight tip, fluted on the inside making it possible to grip sensitive materials reliably.
Broad tip shape: These disposable forceps have ends with a broad tip. The broad tip is smooth and blunt preventing material from being damaged when gripped.
Certifications: Complies with EU food requirements and FDA regulations.
Balení: Available either in a large pack with 100 pieces or 100 pieces packed individually and radiation sterilised.
Specifikace
  • Balené:
    Large pack
  • Sterilní:
    -
  • Type:
    Broad tip, straight
  • Sekvence:
    05