[EN]MICRO TEST PLATE 96-WELL F-BOTTOM PS 1 * 100 KS
RATI6018718
: RATIOLAB
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
Funnel with ball and lid, ARNOLD, V2.0
: S.C.A.T.
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Roztěrky, tvar L, jednorázové
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF