[EN]TUBE CENTRIFUGE 50ML SELF-STANDING 1 * 250 KS
RATI3300003
: RATIOLAB
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Premium Line, Pipety sérologické
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Obličejové štíty, Clearways
: HONEYWELL SAFETY
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Kyselina dusičná 67%, NORMATOM® for trace metal analysis
: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection