[EN]PLATE HT 96W 100µL 2MM GAP UNCOATED 1 * 1 KS
Catalog # BTXI45-0450-M
Supplier: BTX
Frequently Bought Together
Cell culture storage bottles, square, PC, Corning®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Petri dishes, Corning® Gosselin™
Supplier: Corning
VWR®, High Performance Blender Bags
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR®, Kónické nádobky na vzorky, se zaklapávacím víčkem
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Kryoboxy, 133×133 mm
Supplier: TENAK
VWR®, Cryoboxes, PP, Flat-Packed
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Kryoboxy, lepenka, 133×133 mm
Supplier: VWR Collection
Accessories and consumables for KingFisher™ Flex
Supplier: Thermo Fisher Scientific