Fibre optic ring lights
03, Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 86 mm, ext.Ø 114 mm, 18 - 70 mm working distance
Catalog # SCOC157086
Supplier: SCHOTT AG Lighting and imaging
Jump to:
Product Details & Documents
Fibre optic ring lights are available as six point, annular, or darkfield versions with specific advantages. For all ring lights, adapters are available for a variety of objective series. Ring lights are used to provide intense, uniform, and shadow-free illumination.
- Mounts directly to the objective using a thumb screw
- Housing made of rugged, black, anodised aluminium
- Fibre bundle protected by flexible metal PVC sheathing
- High white light transmission thanks to the use of SCHOTT PURAVIS ® fibres
Annular ring lights offer absolute shadow-free illumination. Six point ring lights are a cost effective alternative to annular ring lights. Darkfield ring lights offer low angle illumination for shiny, reflective surfaces.
Ordering information: Cold light source must be ordered separately.
Delivery information: Supplied without cold light source.
Documents
KL Series Fiber Optic Illumination for Stereo Microscopy
Product Literature
TDS Ringlights
Datasheet
Specifications
- Popis:Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 86 mm, ext.Ø 114 mm, 18 - 70 mm working distance
- Sekvence:03