Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Objectives for microscopes, Panthera TEC POL series
Objectives for microscopes, Panthera TEC POL series
02, CCIS® Plan achromatic strain-free objective UC PL P 10×/0,25 (WD=17,4 mm)
Katalogové číslo MOTI1101001707201
Dodavatel:   MOTIC
Objectives for microscopes, Panthera TEC POL series
Katalogové číslo MOTI1101001707201
Dodavatel:  MOTIC
Objectives for microscopes, Panthera TEC POL series
Objectives for microscopes, Panthera TEC POL series
02, CCIS® Plan achromatic strain-free objective UC PL P 10×/0,25 (WD=17,4 mm)
Přejít na:
Price
5 050,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    CCIS® Plan achromatic strain-free objective UC PL P 10×/0,25 (WD=17,4 mm)
  • Sekvence:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.