Accessories for Eppendorf conical tubes
Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal tool
Catalog # EPPE5365000.109
Supplier: EPPENDORF
Accessories for Eppendorf conical tubes
Catalog # EPPE5365000.109
Supplier: EPPENDORF
Jump to:
Product Details & Documents
Specifications
- Popis:Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal tool
- Cat. No.:EPPE5365000.109
- Certifikát ochrany životního prostředí:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact