Orders LinkContactUsLinkComponent
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
12, ESC swab Mycoplasma Transport Broth, 100 tubes×5 ml
Catalog # 85612.
Supplier:   LIOFILCHEM
ESC (Easy Surface Checking) swabs
Catalog # 85612.
Supplier:  LIOFILCHEM
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
12, ESC swab Mycoplasma Transport Broth, 100 tubes×5 ml
Přejít na:
Price
2 900,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Specifikace
  • Popis:
    ESC swab Mycoplasma Transport Broth, 100 tubes×5 ml
  • Sekvence:
    12