[EN]TRANSFER PIPET 50µL CAPILLARY N-ST 1 * 5.000 KS
SAMC1029-100
: SAMCO
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
: VWR Collection
VWR®, Sérologická pipeta, standardní
: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Ubrousky čisticí
: VWR Collection