Objectives for Rebel Microscopes
16, Planachromatický, CSC 0,17, N.A. 0,40, 20×, 1,6 mm
Catalog # ECHLRMAG0202
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Zvětšení:20×
- Popis:Planachromatický, CSC 0,17, N.A. 0,40
- Sekvence:16
- Pracovní vzdálenost:1,6 mm