Objectives for Rebel Microscopes
30, Planachromatický, CSC 0,17, N.A. 0,13, 4×, 17 mm
Catalog # ECHLRMAG0042
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Zvětšení:4×
- Popis:Planachromatický, CSC 0,17, N.A. 0,13
- Sekvence:30
- Pracovní vzdálenost:17 mm