Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
01, #1, 3 mm
Catalog # HECH41001103
Cover glasses
Catalog # HECH41001103
Cover glasses
Cover glasses
01, #1, 3 mm
Price
14 010,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
Ordering information: Available in different diameters.
Specifications
  • Ø:
    3 mm
  • Počet:
    #1
  • Sekvence:
    01