HIGH SELECT HSA IMMUNOGLOBULIN MINI SPIN 1 * 6 KS
PIERA36365
Frequently Bought Together
Serological pipettes, Stripette™
: Corning
VWR®, Zátky, PE
: VWR Collection
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
VWR®, Roztěrky, tvar L, jednorázové
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
terc-Butylmethylether ≥99%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Misky kultivační, vícejamkové
: VWR Collection
Lab blender bags, Stomacher®
: Seward (FermionX)
Destička J.T.Baker® SRW, magnetická
: Avantor
Dichlormethan ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sterilised write-on bags, Whirl-Pak®
: NASCO FORT ATKINSON
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection