[EN]Methanol UHPLC for mass spectrometry 1 * 1 L
Katalogové číslo SIAL900688-1L
Dodavatel: Merck
Frequently Bought Together
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Voda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Dodavatel: Merck
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Voda USP, JP, Ph. Eur., purified
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Toluen ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Petrolether, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck