Labels for BradyJet J2000 Lab printer
22, Ink pads, J20-PAD, Inkjet, Bílá, J2000, 127×3 mm
Katalogové číslo BRDY150163
Dodavatel: Brady
Přejít na:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Brady Jet J2000 Laboratory Colour Label Printer
Produktová literatura
Specifikace
- Printer model:J2000
- Printing method:Inkjet
- Barva:Bílá
- Popis:Ink pads
- Rozměry:127×3 mm
- Sekvence:22
- Model:J20-PAD