Orders LinkContactUsLinkComponent
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
Reading magnifier, black, visoPOCKET, 2,5×, 74×48 mm
Katalogové číslo ESCH172156
Dodavatel:   ESCHENBACH OPTIK
Reading magnifiers, visoPOCKET
Katalogové číslo ESCH172156
Dodavatel:  ESCHENBACH OPTIK
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
Reading magnifier, black, visoPOCKET, 2,5×, 74×48 mm
Přejít na:
Price
920,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Compact magnifiers with wide field of view and ultra-slim lenses.
  • Diffractive aspheric lens with modern and exquisite design
  • High quality leather available in three colours
  • Leather case protects from scratches
Sutiable for on-the-go use when small print needs magnifying swiftly and easily.
Specifikace
  • Zvětšení:
    2,5×
  • Popis:
    Reading magnifier, black, visoPOCKET
  • Š×H:
    74×48 mm