Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
18, 2-way valve system, 19 mm, 2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Katalogové číslo HUBE350025
Dodavatel:   HUBER
Accessories for circulators
Katalogové číslo HUBE350025
Dodavatel:  HUBER
Accessories for circulators
18, 2-way valve system, 19 mm, 2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Přejít na:
Price
14 880,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    2-way valve system, 19 mm
  • Sekvence:
    18
  • Určeno pro:
    2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.