[EN]EJECTION INTERFACE MULTI L, L AUTO 1 * 1 KS
Katalogové číslo GILSFA07097
Dodavatel: GILSON
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
epT.I.P.S.® Racks - Pipette Tips
Dodavatel: EPPENDORF
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Plynové kartuše, Campingaz®
Dodavatel: CAMPING GAZ
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Příslušenství pro mechanické pipety
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Dodavatel: HIRSCHMANN