Orders LinkContactUsLinkComponent
Sponge sticks
Sponge sticks
04, Media type: Letheen broth
Katalogové číslo MMMPSSL10LET
Dodavatel:   Neogen
Sponge sticks
Katalogové číslo MMMPSSL10LET
Dodavatel:  Neogen
Sponge sticks
Sponge sticks
04, Media type: Letheen broth
Přejít na:
Price
7 600,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
3M™ Sponge-Stick in sample bag with 10 ml D/E neutralising broth uses simplified packaging to minimise handling and preparation, and reduces waste.
  • Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge
  • Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing
  • Simplified packaging minimises handling and preparation and reduces waste
Better testing begins with better samples and better samples are obtained when you use the best collection tools. 3M™ Sponge-Stick allows sampling without directly handling the sponge. It makes it easier to reach into drains, pipes and around equipment. Biocide-free cellulose sponge maintains organism viability. These are convenient and ready-to-use with consistent, reliable results every time.
Informace o dodávce: Supplied in wet or dry formats to fit many applications.
Documents
Specifikace
  • Popis:
    Media type: Letheen broth
  • Sekvence:
    04