Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
VWR® Hichrom Nuts and Ferrules
VWR® Hichrom Nuts and Ferrules
06, 1/16" PEEK fingertight ferrule
Katalogové číslo HI-508
Dodavatel:   Avantor
VWR® Hichrom Nuts and Ferrules
Katalogové číslo HI-508
Dodavatel:  Avantor
VWR® Hichrom Nuts and Ferrules
VWR® Hichrom Nuts and Ferrules
06, 1/16" PEEK fingertight ferrule
Přejít na:
Price
2 860,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Precision-machined stainless steel nuts and ferrules.
  • Burr and contaminant free
Specifikace
  • Popis:
    1/16" PEEK fingertight ferrule
  • Sekvence:
    06
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.