[EN]DROPETTE N/STE. 4ML 155MM(L) BULB 3. 1 * 500 KS
SIMPP200-44
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Vzorkovací lžíce
: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
: VWR Collection