[EN]TESTING THE ENVIRONMENTAL IMPACTS OF 1 * 1 KIT
EDVO905
: EDVOTEK
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Voda, TECHNICAL, demineralised
: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals